Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Salicin, MP Biomedicals

Catalog No. ICN10283380 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10283380 100 g
ICN10283305 5 g
ICN10283310 10 g
ICN10283325 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN10283380 Supplier MP Biomedicals, Inc Supplier No. 0210283380
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Salicin is a substrate for β-glucosidase

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138-52-3
Molecular Formula C13H18O7
Molecular Weight (g/mol) 286.28
MDL Number MFCD00006590
InChI Key NGFMICBWJRZIBI-UJPOAAIJSA-N
Synonym salicin, salicoside, salicine, salicyl alcohol glucoside, d---salicin, 2r,3s,4s,5r,6s-2-hydroxymethyl-6-2-hydroxymethyl phenoxy tetrahydro-2h-pyran-3,4,5-triol, saligenin beta-d-glucopyranoside, 2-hydroxymethyl phenyl beta-d-glucopyranoside, d-salicin, saligenin-beta-d-glucopyranoside
PubChem CID 439503
ChEBI CHEBI:17814
IUPAC Name (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-[2-(hydroxymethyl)phenoxy]oxane-3,4,5-triol
SMILES C1=CC=C(C(=C1)CO)OC2C(C(C(C(O2)CO)O)O)O

Specifications

Quantity 100 g
Formula Weight 286.3
Grade Research
Physical Form Powder
Chemical Name or Material Salicin
EINECSNumber 205-331-6
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.