accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHCH3HO

CAS RN 138090-06-9

IUPAC Name: 5,11-diethyl-5,6,11,12-tetrahydrochrysene-2,8-diol
Synonyms: (R,R)-tetrahydrochrysene
Molecular Weight (g/mol): 320.43
Molecular Formula: C22H24O2
InChi Key: MASYAWHPJCQLSW-UHFFFAOYNA-N
SMILES: CCC1CC2=C(C=CC(O)=C2)C2=C1C1=C(CC2CC)C=C(O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results