accessibility menu, dialog, popup

UserName

Viewing profile as user:
OSO2HOO2H

CAS RN 13813-19-9

IUPAC Name: (²H₂)sulfuric acid
Synonyms: (²H₂)sulfuric acid
Molecular Weight (g/mol): 100.08
Molecular Formula: H2O4S
InChi Key: QAOWNCQODCNURD-ZSJDYOACSA-N
SMILES: [2H]OS(=O)(=O)O[2H]
Quantity 
  • (1)
  • (1)
Grade 
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results