Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Deuterosulfuric acid, 96% min in D{2}O, 99.5% (Isotopic)

Catalog No. AA8785718
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA8785718 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA8785718 Supplier Thermo Scientific Chemicals Supplier No. 08785718
Add to Cart

CAS: 13813-19-9 | H2O4S | 100.08 g/mol

Deuterated sulfuric acid when treated with benzene, the deuterium is used in order to observe the reaction course by NMR spectroscopy, deuterated sulfuric acid results in an electrophilic aromatic substitution of hydrogen by deuterium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13813-19-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 100.08
MDL Number MFCD00003542
InChI Key QAOWNCQODCNURD-ZSJDYOACSA-N
Synonym sulfuric acid-d2, deuterosulfuric acid, dideuterosulfuric acid, sulphuric 2h2 acid, sulfuric acid-d2 solution, d2so4, dideuterosulfuric acid solution, 2 h? sulfuric acid, sulfuric acid-d2, 98 wt% in deuterium oxide, deuterosulfuric acid min in d 2 o isotopic
PubChem CID 160992
IUPAC Name dideuterio sulfate
SMILES [2H]OS(=O)(=O)O[2H]

Specifications

Physical Form Liquid
Sensitivity Moisture
Quantity 50 g
Refractive Index 1.4175
Density 1.86
Boiling Point 290°C
Assay Percent Range <96% in D2O, 99.5% (Isotopic)
Solubility Information Soluble in water
Formula Weight 100.09
Chemical Name or Material Deuterosulfuric Acid
Hazard Category H314-H335
Hazard Statement In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Never add water to this product. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 237-464-0
TSCA Yes
Recommended Storage Ambient

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.