Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric acid-d2, for NMR, 98 wt.% in D2O, 99.5+ atom % D

Catalog No. AC175481000
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC175481000 100 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC175481000 Supplier Thermo Scientific Chemicals Supplier No. 175481000
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 13813-19-9 | H2O4S | 100.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13813-19-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 100.08
MDL Number MFCD00003542
InChI Key QAOWNCQODCNURD-ZSJDYOACSA-N
Synonym sulfuric acid-d2, deuterosulfuric acid, dideuterosulfuric acid, sulphuric 2h2 acid, sulfuric acid-d2 solution, d2so4, dideuterosulfuric acid solution, 2 h? sulfuric acid, sulfuric acid-d2, 98 wt% in deuterium oxide, deuterosulfuric acid min in d 2 o isotopic
PubChem CID 160992
IUPAC Name dideuterio sulfate
SMILES [2H]OS(=O)(=O)O[2H]

Specifications

Physical Form Liquid
Color Colorless
Packaging Glass Bottle
Quantity 100 mL
Deuterium (D) 99.50% min.
Density 1.8700g/mL
Linear Formula D2SO4
Formula Weight 100.09
Percent Purity 97.5 to 102.5%
Grade NMR
Chemical Name or Material Sulfuric acid-d2, 99.5+ atom % D
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 237-464-0
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.