accessibility menu, dialog, popup

UserName

Viewing profile as user:
IIIEr

CAS RN 13813-42-8

IUPAC Name: erbium(3+) triiodide
Synonyms: erbium(III) iodide
Molecular Weight (g/mol): 547.97
Molecular Formula: ErI3
InChi Key: OKVQKDALNLHZLB-UHFFFAOYSA-K
SMILES: [I-].[I-].[I-].[Er+3]
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)