Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Erbium(III) iodide, anhydrous, REacton™, 99.99% (REO)

Catalog No. AA4337218
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4337218 50 g
AA4337209 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4337218 Supplier Thermo Scientific Chemicals Supplier No. 04337218
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 13813-42-8 | ErI3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13813-42-8
Molecular Formula ErI3
MDL Number MFCD00016075
Synonym erbium iii iodide, erbium iii ; iodide, acmc-1bohg, erbium iodide,

Specifications

Boiling Point 1280°C
Melting Point 1020°C
Physical Form Powder
Quantity 50 g
Formula Weight 547.97
Percent Purity 99.99%
Grade Reacton
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Erbium (III) iodide, Anhydrous
EINECSNumber 237-471-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.