Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Erbium(III) iodide, ultra dry, 99.9% (REO)

Catalog No. AA3569506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3569506 5 g
AA3569503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3569506 Supplier Thermo Scientific Chemicals Supplier No. 03569506
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 13813-42-8

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13813-42-8
MDL Number MFCD00016075
Synonym erbium iii iodide, erbium iii ; iodide, acmc-1bohg, erbium iodide,

Specifications

Boiling Point 1280°C
Melting Point 1020°C
Physical Form Powder or Pieces
Quantity 5 g
Formula Weight 547.97
Percent Purity 99.9%
Packaging Ampule
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Erbium(III) iodide, Ultra dry
EINECSNumber 237-471-9
TSCA Yes
storageNote1
  • Ampouled under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.