accessibility menu, dialog, popup

UserName

Viewing profile as user:
NClNCl

CAS RN 138219-98-4

IUPAC Name: 4,4'-bis(chloromethyl)-2,2'-bipyridine
Synonyms: 4,4'-bis(chloromethyl)-2,2'-bipyridine
Molecular Weight (g/mol): 253.13
Molecular Formula: C12H10Cl2N2
InChi Key: VZKSOWFFOHQARA-UHFFFAOYSA-N
SMILES: ClCC1=CC(=NC=C1)C1=NC=CC(CCl)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results