accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1383478-94-1

IUPAC Name: N-[4-(dimethylsulfamoyl)phenyl]-4-(hydroxydiphenylmethyl)piperidine-1-carbothioamide
Synonyms: N-[4-(dimethylsulfamoyl)phenyl]-4-(hydroxydiphenylmethyl)piperidine-1-carbothioamide
Molecular Weight (g/mol): 509.68
Molecular Formula: C27H31N3O3S2
InChi Key: VAUKHARIAMDAGF-UHFFFAOYSA-N
SMILES: CN(C)S(=O)(=O)C1=CC=C(NC(=S)N2CCC(CC2)C(O)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results