CAS RN 138356-20-4
IUPAC Name:
[2-(3,4-dichlorophenyl)ethyl][2-(dimethylamino)ethyl]methylamine
Synonyms:
[2-(3,4-dichlorophenyl)ethyl][2-(dimethylamino)ethyl]methylamine
Molecular Weight (g/mol):
275.22
Molecular Formula:
C13H20Cl2N2
InChi Key:
MGVRNMUKTZOQOW-UHFFFAOYSA-N
SMILES:
CN(C)CCN(C)CCC1=CC=C(Cl)C(Cl)=C1
1
–
1
of
1
results
1
Tocris Bioscience™ BD 1047 dihydrobromide
CAS: 138356-20-4 Molecular Formula: C13H20Cl2N2