accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNONHONSHNNH2CH3SOOHO

CAS RN 138489-18-6

IUPAC Name: [(3-{3-[4-(1-methyl-1H-indol-3-yl)-2,5-dioxo-2,5-dihydro-1H-pyrrol-3-yl]-1H-indol-1-yl}propyl)sulfanyl]methanimidamide; methanesulfonic acid
Synonyms: (3-{3-[4-(1-methylindol-3-yl)-2,5-dioxo-1H-pyrrol-3-yl]indol-1-yl}propyl)sulfanylmethanimidamide; methanesulfonic acid
Molecular Weight (g/mol): 553.65
Molecular Formula: C26H27N5O5S2
InChi Key: SAWVGDJBSPLRRB-UHFFFAOYSA-N
SMILES: CS(O)(=O)=O.CN1C=C(C2=CC=CC=C12)C1=C(C(=O)NC1=O)C1=CN(CCCSC(N)=N)C2=CC=CC=C12
Form 
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results