accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OONHF

CAS RN 1385571-81-2

IUPAC Name: N-[(3-fluorophenyl)methyl]-2-methoxybenzamide
Synonyms: N-[(3-fluorophenyl)methyl]-2-methoxybenzamide
Molecular Weight (g/mol): 259.28
Molecular Formula: C15H14FNO2
InChi Key: BHXOHPQLWQFDDN-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1C(=O)NCC1=CC(F)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results