accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3OONCH3OOHO

CAS RN 138774-98-8

IUPAC Name: 3-[4-(benzyloxy)phenyl]-2-{[(tert-butoxy)carbonyl](methyl)amino}propanoic acid
Synonyms: 3-[4-(benzyloxy)phenyl]-2-[(tert-butoxycarbonyl)(methyl)amino]propanoic acid
Molecular Weight (g/mol): 385.46
Molecular Formula: C22H27NO5
InChi Key: FSNRGORPOYPIJC-UHFFFAOYNA-N
SMILES: CN(C(CC1=CC=C(OCC2=CC=CC=C2)C=C1)C(O)=O)C(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)