accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOOOOHNOO

CAS RN 138906-43-1

IUPAC Name: 2-[8-hydroxy-6-(4-methoxyphenoxy)-2-phenyl-hexahydro-2H-pyrano[3,2-d][1,3]dioxin-7-yl]-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: 2-[8-hydroxy-6-(4-methoxyphenoxy)-2-phenyl-hexahydro-2H-pyrano[3,2-d][1,3]dioxin-7-yl]isoindole-1,3-dione
Molecular Weight (g/mol): 503.51
Molecular Formula: C28H25NO8
InChi Key: SHSWKRFHEJOKLK-UHFFFAOYNA-N
SMILES: COC1=CC=C(OC2OC3COC(OC3C(O)C2N2C(=O)C3=CC=CC=C3C2=O)C2=CC=CC=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results