accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2SH2N

CAS RN 139-65-1

IUPAC Name: 4-[(4-aminophenyl)sulfanyl]aniline
Synonyms: 4,4'-thiodianiline
Molecular Weight (g/mol): 216.3
Molecular Formula: C12H12N2S
InChi Key: ICNFHJVPAJKPHW-UHFFFAOYSA-N
SMILES: NC1=CC=C(SC2=CC=C(N)C=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results