accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NNHOHNNNNO

CAS RN 139180-30-6

IUPAC Name: 4-(2-{[7-amino-2-(furan-2-yl)-[1,2,4]triazolo[1,5-a][1,3,5]triazin-5-yl]amino}ethyl)phenol
Synonyms: 4-(2-{[7-amino-2-(furan-2-yl)-[1,2,4]triazolo[1,5-a][1,3,5]triazin-5-yl]amino}ethyl)phenol
Molecular Weight (g/mol): 337.34
Molecular Formula: C16H15N7O2
InChi Key: PWTBZOIUWZOPFT-UHFFFAOYSA-N
SMILES: NC1=NC(NCCC2=CC=C(O)C=C2)=NC2=NC(=NN12)C1=CC=CO1
Form 
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results