accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOHNFOCH3

CAS RN 139290-65-6

IUPAC Name: (2,3-dimethoxyphenyl)({1-[2-(4-fluorophenyl)ethyl]piperidin-4-yl})methanol
Synonyms: volinanserin
Molecular Weight (g/mol): 373.47
Molecular Formula: C22H28FNO3
InChi Key: HXTGXYRHXAGCFP-UHFFFAOYNA-N
SMILES: COC1=CC=CC(C(O)C2CCN(CCC3=CC=C(F)C=C3)CC2)=C1OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results