accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1393814-38-4

IUPAC Name: 5-(3-chloro-4-cyclohexylphenyl)-1-(3-methoxyphenyl)-1H-pyrazole-3-carboxylic acid
Synonyms: 5-(3-chloro-4-cyclohexylphenyl)-1-(3-methoxyphenyl)pyrazole-3-carboxylic acid
Molecular Weight (g/mol): 410.9
Molecular Formula: C23H23ClN2O3
InChi Key: BNALUYKEGYUHQC-UHFFFAOYSA-N
SMILES: COC1=CC=CC(=C1)N1N=C(C=C1C1=CC(Cl)=C(C=C1)C1CCCCC1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results