accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOIOOHOOONaNaNa

CAS RN 13940-38-0

IUPAC Name: trisodium (dihydroxydioxidoiodosyl)olate
Synonyms: trisodium orthoperiodate(3-)
Molecular Weight (g/mol): 293.88
Molecular Formula: H2INa3O6
InChi Key: DSYMXPNUZLZKOA-UHFFFAOYSA-K
SMILES: [Na+].[Na+].[Na+].O[I](O)([O-])([O-])([O-])=O
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)