accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1396006-71-5

IUPAC Name: 1-[4'-(4-methyl-5-{[(1-phenylethoxy)carbonyl]amino}-1H-1,2,3-triazol-1-yl)-[1,1'-biphenyl]-4-yl]cyclopropane-1-carboxylic acid
Synonyms: 1-[4'-(4-methyl-5-{[(1-phenylethoxy)carbonyl]amino}-1,2,3-triazol-1-yl)-[1,1'-biphenyl]-4-yl]cyclopropane-1-carboxylic acid
Molecular Weight (g/mol): 482.54
Molecular Formula: C28H26N4O4
InChi Key: PXQUHYSYFWQRMF-UHFFFAOYNA-N
SMILES: CC(OC(=O)NC1=C(C)N=NN1C1=CC=C(C=C1)C1=CC=C(C=C1)C1(CC1)C(O)=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results