accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOH3CPOHOHNClCl

CAS RN 139667-74-6

IUPAC Name: (3-{[(3,4-dichlorophenyl)methyl]amino}propyl)(diethoxymethyl)phosphinic acid
Synonyms: 3-{[(3,4-dichlorophenyl)methyl]amino}propyl(diethoxymethyl)phosphinic acid
Molecular Weight (g/mol): 384.23
Molecular Formula: C15H24Cl2NO4P
InChi Key: GJZVQXWEIYRHBE-UHFFFAOYNA-N
SMILES: CCOC(OCC)P(O)(=O)CCCNCC1=CC=C(Cl)C(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)