accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3(R)O(R)O(R)(S)O(R)HO(S)HO(R)HO(R)HO(R)OH(R)OHOO(S)CH3(R)CH3(R)OH(S)CH3(S)OH(R)HOO(S)HO(S)H2N(S)OH(?)(?)(?)(?)(?)(?)(?)

CAS RN 1397-89-3

IUPAC Name: (1R,3S,5R,6R,9R,11R,15S,16R,17R,18S,19Z,21Z,23Z,25Z,27Z,29Z,31Z,33R,35S,36R,37S)-33-{[(2R,3S,4S,5S,6R)-4-amino-3,5-dihydroxy-6-methyloxan-2-yl]oxy}-1,3,5,6,9,11,17,37-octahydroxy-15,16,18-trimethyl-13-oxo-14,39-dioxabicyclo[33.3.1]nonatriaconta-19,21,23,25,27,29,31-heptaene-36-carboxylic acid
Synonyms: amphotericin B
Molecular Weight (g/mol): 924.09
Molecular Formula: C47H73NO17
InChi Key: APKFDSVGJQXUKY-ZNVUZQDLSA-N
SMILES: C[C@H]1O[C@@H](O[C@@H]2C[C@@H]3O[C@@](O)(C[C@H](O)[C@H]3C(O)=O)C[C@@H](O)C[C@@H](O)[C@H](O)CC[C@@H](O)C[C@@H](O)CC(=O)O[C@@H](C)[C@H](C)[C@H](O)[C@@H](C)\C=C/C=C\C=C/C=C\C=C/C=C\C=C/2)[C@@H](O)[C@@H](N)[C@@H]1O
Quantity 
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (4)
  • (3)
  • (1)
  • (1)
Type 
  • (5)
  • (17)
Physical Form 
  • (5)
  • (13)
  • (1)
Color 
  • (5)
  • (12)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (3)
  • (5)
  • (1)
  • (7)
  • (7)
  • (1)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (4)
  • (3)
  • (1)
  • (1)
  • (5)
  • (17)
  • (5)
  • (13)
  • (1)
  • (5)
  • (12)
  • (1)
  • (1)
  • (7)
  • (4)
  • (5)
  • (2)
  • (1)