accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OH3CCH3H3COSOOCl

CAS RN 140-57-8

IUPAC Name: 1-(4-tert-butylphenoxy)propan-2-yl 2-chloroethyl sulfite
Synonyms: aramite
Molecular Weight (g/mol): 334.86
Molecular Formula: C15H23ClO4S
InChi Key: YKFRAOGHWKADFJ-UHFFFAOYNA-N
SMILES: CC(COC1=CC=C(C=C1)C(C)(C)C)OS(=O)OCCCl
Concentration 
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)