accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOOH

CAS RN 14003-96-4

IUPAC Name: 7-hydroxy-4-methyl-2-oxo-2H-chromene-8-carbaldehyde
Synonyms: 7-hydroxy-4-methyl-2-oxochromene-8-carbaldehyde
Molecular Weight (g/mol): 204.18
Molecular Formula: C11H8O4
InChi Key: RTHHSXOVIJWFQP-UHFFFAOYSA-N
SMILES: CC1=CC(=O)OC2=C(C=O)C(O)=CC=C12
Form 
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results