CAS RN 1401-55-4
Tannic acid, ACS
CAS: 1401-55-4 Molecular Formula: C76H52O46 Molecular Weight (g/mol): 1701.206 MDL Number: MFCD00066397 InChI Key: LRBQNJMCXXYXIU-JPZKJCTESA-N Synonym: tannins,gallotannic acid,quebracho extract,acide tannique,d'acide tannique,tannic acid usp:jan,unii-28f9e0djy6 PubChem CID: 134129492 SMILES: C1=C(C=C(C(=C1O)O)O)C(=O)OC2=CC(=CC(=C2O)O)C(=O)OCC3C(C(C(C(O3)OC(=O)C4=CC(=C(C(=C4)OC(=O)C5=CC(=C(C(=C5)O)O)O)O)O)OC(=O)C6=CC(=C(C(=C6)OC(=O)C7=CC(=C(C(=C7)O)O)O)O)O)OC(=O)C8=CC(=C(C(=C8)OC(=O)C9=CC(=C(C(=C9)O)O)O)O)O)OC(=O)C1=CC(=C(C(=C1)OC(=O)C1=CC(=C(
Tannic acid, 95%
CAS: 1401-55-4 Molecular Formula: C76H52O46 Molecular Weight (g/mol): 1701.206 MDL Number: MFCD00066397 InChI Key: LRBQNJMCXXYXIU-JPZKJCTESA-N Synonym: tannins,gallotannic acid,quebracho extract,acide tannique,d'acide tannique,tannic acid usp:jan,unii-28f9e0djy6 PubChem CID: 134129492 SMILES: C1=C(C=C(C(=C1O)O)O)C(=O)OC2=CC(=CC(=C2O)O)C(=O)OCC3C(C(C(C(O3)OC(=O)C4=CC(=C(C(=C4)OC(=O)C5=CC(=C(C(=C5)O)O)O)O)O)OC(=O)C6=CC(=C(C(=C6)OC(=O)C7=CC(=C(C(=C7)O)O)O)O)O)OC(=O)C8=CC(=C(C(=C8)OC(=O)C9=CC(=C(C(=C9)O)O)O)O)O)OC(=O)C1=CC(=C(C(=C1)OC(=O)C1=CC(=C(
Tannic Acid, USP, Spectrum™ Chemical
CAS: 1401-55-4 Molecular Formula: C76H52O46 Molecular Weight (g/mol): 1701.21 InChI Key: LRBQNJMCXXYXIU-UHFFFAOYNA-N IUPAC Name: 2,3-dihydroxy-5-[({3,4,5,6-tetrakis[3,4-dihydroxy-5-(3,4,5-trihydroxybenzoyloxy)benzoyloxy]oxan-2-yl}methoxy)carbonyl]phenyl 3,4,5-trihydroxybenzoate SMILES: OC1=CC(=CC(O)=C1O)C(=O)OC1=CC(=CC(O)=C1O)C(=O)OCC1OC(OC(=O)C2=CC(O)=C(O)C(OC(=O)C3=CC(O)=C(O)C(O)=C3)=C2)C(OC(=O)C2=CC(O)=C(O)C(OC(=O)C3=CC(O)=C(O)C(O)=C3)=C2)C(OC(=O)C2=CC(O)=C(O)C(OC(=O)C3=CC(O)=C(O)C(O)=C3)=C2)C1OC(=O)C1=CC(O)=C(O)C(OC(=O)C2=CC(O)=C(O)C(O)=C2)=C1
Tannic Acid, USP, Spectrum™ Chemical
CAS: 1401-55-4 Molecular Formula: C76H52O46 Molecular Weight (g/mol): 1701.21 InChI Key: LRBQNJMCXXYXIU-UHFFFAOYNA-N IUPAC Name: 2,3-dihydroxy-5-[({3,4,5,6-tetrakis[3,4-dihydroxy-5-(3,4,5-trihydroxybenzoyloxy)benzoyloxy]oxan-2-yl}methoxy)carbonyl]phenyl 3,4,5-trihydroxybenzoate SMILES: OC1=CC(=CC(O)=C1O)C(=O)OC1=CC(=CC(O)=C1O)C(=O)OCC1OC(OC(=O)C2=CC(O)=C(O)C(OC(=O)C3=CC(O)=C(O)C(O)=C3)=C2)C(OC(=O)C2=CC(O)=C(O)C(OC(=O)C3=CC(O)=C(O)C(O)=C3)=C2)C(OC(=O)C2=CC(O)=C(O)C(OC(=O)C3=CC(O)=C(O)C(O)=C3)=C2)C1OC(=O)C1=CC(O)=C(O)C(OC(=O)C2=CC(O)=C(O)C(O)=C2)=C1
Tannic acid, ACS reagent
CAS: 1401-55-4 Molecular Formula: C76H52O46 Molecular Weight (g/mol): 1701.206 MDL Number: MFCD00066397 InChI Key: LRBQNJMCXXYXIU-JPZKJCTESA-N Synonym: tannins,gallotannic acid,quebracho extract,acide tannique,d'acide tannique,tannic acid usp:jan,unii-28f9e0djy6 PubChem CID: 134129492 SMILES: C1=C(C=C(C(=C1O)O)O)C(=O)OC2=CC(=CC(=C2O)O)C(=O)OCC3C(C(C(C(O3)OC(=O)C4=CC(=C(C(=C4)OC(=O)C5=CC(=C(C(=C5)O)O)O)O)O)OC(=O)C6=CC(=C(C(=C6)OC(=O)C7=CC(=C(C(=C7)O)O)O)O)O)OC(=O)C8=CC(=C(C(=C8)OC(=O)C9=CC(=C(C(=C9)O)O)O)O)O)OC(=O)C1=CC(=C(C(=C1)OC(=O)C1=CC(=C(
Tannic acid
CAS: 1401-55-4 Molecular Formula: C76H52O46 Molecular Weight (g/mol): 1701.206 MDL Number: MFCD00066397 InChI Key: LRBQNJMCXXYXIU-JPZKJCTESA-N Synonym: tannins,gallotannic acid,quebracho extract,acide tannique,d'acide tannique,tannic acid usp:jan,unii-28f9e0djy6 PubChem CID: 134129492 SMILES: C1=C(C=C(C(=C1O)O)O)C(=O)OC2=CC(=CC(=C2O)O)C(=O)OCC3C(C(C(C(O3)OC(=O)C4=CC(=C(C(=C4)OC(=O)C5=CC(=C(C(=C5)O)O)O)O)O)OC(=O)C6=CC(=C(C(=C6)OC(=O)C7=CC(=C(C(=C7)O)O)O)O)O)OC(=O)C8=CC(=C(C(=C8)OC(=O)C9=CC(=C(C(=C9)O)O)O)O)O)OC(=O)C1=CC(=C(C(=C1)OC(=O)C1=CC(=C(
Tannic Acid, Reagents
CAS: 1401-55-4 Molecular Formula: C76H52O46 Molecular Weight (g/mol): 1701.21 InChI Key: LRBQNJMCXXYXIU-UHFFFAOYNA-N Synonym: Gallotannic Acid, Glycerite IUPAC Name: 2,3-dihydroxy-5-[({3,4,5,6-tetrakis[3,4-dihydroxy-5-(3,4,5-trihydroxybenzoyloxy)benzoyloxy]oxan-2-yl}methoxy)carbonyl]phenyl 3,4,5-trihydroxybenzoate SMILES: OC1=CC(=CC(O)=C1O)C(=O)OC1=CC(=CC(O)=C1O)C(=O)OCC1OC(OC(=O)C2=CC(O)=C(O)C(OC(=O)C3=CC(O)=C(O)C(O)=C3)=C2)C(OC(=O)C2=CC(O)=C(O)C(OC(=O)C3=CC(O)=C(O)C(O)=C3)=C2)C(OC(=O)C2=CC(O)=C(O)C(OC(=O)C3=CC(O)=C(O)C(O)=C3)=C2)C1OC(=O)C1=CC(O)=C(O)C(OC(=O)C2=CC(O)=C(O)C(O)=C2)=C1
Tannic Acid, Solstice
CAS: 1401-55-4 Molecular Formula: C76H52O46 Molecular Weight (g/mol): 1701.206 MDL Number: MFCD00066397 InChI Key: LRBQNJMCXXYXIU-JPZKJCTESA-N Synonym: tannins,gallotannic acid,quebracho extract,acide tannique,d'acide tannique,tannic acid usp:jan,unii-28f9e0djy6 PubChem CID: 134129492 SMILES: C1=C(C=C(C(=C1O)O)O)C(=O)OC2=CC(=CC(=C2O)O)C(=O)OCC3C(C(C(C(O3)OC(=O)C4=CC(=C(C(=C4)OC(=O)C5=CC(=C(C(=C5)O)O)O)O)O)OC(=O)C6=CC(=C(C(=C6)OC(=O)C7=CC(=C(C(=C7)O)O)O)O)O)OC(=O)C8=CC(=C(C(=C8)OC(=O)C9=CC(=C(C(=C9)O)O)O)O)O)OC(=O)C1=CC(=C(C(=C1)OC(=O)C1=CC(=C(
Sigma Aldrich [13C]-Silaco
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Tannic Acid
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Tannic Acid
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Fisher Science Education™ Tannic Acid
CAS: 1401-55-4 Molecular Formula: C76H52O46 Molecular Weight (g/mol): 1701.206 InChI Key: LRBQNJMCXXYXIU-JPZKJCTESA-N Synonym: tannins,gallotannic acid,quebracho extract,acide tannique,d'acide tannique,tannic acid usp:jan,unii-28f9e0djy6 PubChem CID: 134129492 SMILES: C1=C(C=C(C(=C1O)O)O)C(=O)OC2=CC(=CC(=C2O)O)C(=O)OCC3C(C(C(C(O3)OC(=O)C4=CC(=C(C(=C4)OC(=O)C5=CC(=C(C(=C5)O)O)O)O)O)OC(=O)C6=CC(=C(C(=C6)OC(=O)C7=CC(=C(C(=C7)O)O)O)O)O)OC(=O)C8=CC(=C(C(=C8)OC(=O)C9=CC(=C(C(=C9)O)O)O)O)O)OC(=O)C1=CC(=C(C(=C1)OC(=O)C1=CC(=C(