Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tannic Acid, Solstice

Catalog No. 60044932 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
25 kg
6 x 1 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-044-932 6 x 1 kg
60-020-20 100 g
60-020-21 1 kg
60-020-23 25 kg
60-020-24 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 60-044-932 Supplier Solstice Supplier No. 162016X1KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Puriss., meets analytical specification of USP, powder

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1401-55-4
Molecular Formula C76H52O46
Molecular Weight (g/mol) 1701.206
MDL Number MFCD00066397
InChI Key LRBQNJMCXXYXIU-JPZKJCTESA-N
Synonym tannins, gallotannic acid, quebracho extract, acide tannique, d'acide tannique, tannic acid usp:jan, unii-28f9e0djy6
PubChem CID 134129492
SMILES C1=C(C=C(C(=C1O)O)O)C(=O)OC2=CC(=CC(=C2O)O)C(=O)OCC3C(C(C(C(O3)OC(=O)C4=CC(=C(C(=C4)OC(=O)C5=CC(=C(C(=C5)O)O)O)O)O)OC(=O)C6=CC(=C(C(=C6)OC(=O)C7=CC(=C(C(=C7)O)O)O)O)O)OC(=O)C8=CC(=C(C(=C8)OC(=O)C9=CC(=C(C(=C9)O)O)O)O)O)OC(=O)C1=CC(=C(C(=C1)OC(=O)C1=CC(=C(

Specifications

Quantity 6 x 1 kg
Flash Point 199°C (390.2°F)
Ignition Point 980°F
Residue after Ignition ≤0.1% (as SO4)
UN Number NONH for all modes of transport
Solubility Information Soluble in acetone, alcohol, water 1gm/0.35mL, glycerol 1gm/1mL (warm). Insoluble in carbon disulfide, chloroform, diethyl ether,carbon tetrachloride, petroleum ether,benzene, hexane. Slightly soluble in dehydrated alcohol.
Formula Weight 1701.2
Grade puriss.
Chemical Name or Material Tannic Acid, Meets analytical specification of USP
Precautionary Statement H412
EINECSNumber 215-753-2
RTECSNumber WW5075000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.