accessibility menu, dialog, popup

UserName

Viewing profile as user:
NBr

CAS RN 1401863-51-1

IUPAC Name: 9-benzyl-2-bromo-9H-carbazole
Synonyms: 9-benzyl-2-bromocarbazole
Molecular Weight (g/mol): 336.23
Molecular Formula: C19H14BrN
InChi Key: BFHNUHBRWXASFQ-UHFFFAOYSA-N
SMILES: BrC1=CC2=C(C=C1)C1=C(C=CC=C1)N2CC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results