CAS RN 14023-80-4
IUPAC Name:
iridium(1+) bis(methanidylidyneoxidanium) (2Z)-4-oxopent-2-en-2-olate
Synonyms:
iridium(1+) bis(carbon monoxide) (2Z)-4-oxopent-2-en-2-olate
Molecular Weight (g/mol):
347.35
Molecular Formula:
C7H7IrO4
InChi Key:
DFIAYCFHPDOZLK-FGSKAQBVSA-M
SMILES:
[Ir+].[C-]#[O+].[C-]#[O+].C\C([O-])=C\C(C)=O
1
–
1
of
1
results
1
Sigma Aldrich Allyltris(3,3,4,4,5,5,6,6,6-Nonafluorohexyl)Stannane
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.