CAS RN 1405-69-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
MP Biomedicals™ Avidin from Egg White
Avidin from Egg White, 10-15 u/mg solid, White Crystalline Powder
2
Thermo Scientific Chemicals Avidin, from egg
CAS: 1405-69-2