accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHONNNNNH2NOH2CH3CH3C

CAS RN 1407966-77-1

IUPAC Name: 3-{4-amino-1-[1-(prop-2-enoyl)piperidin-3-yl]-1H-pyrazolo[3,4-d]pyrimidin-3-yl}-N-[3-methyl-4-(propan-2-yl)phenyl]benzamide
Synonyms: 3-{4-amino-1-[1-(prop-2-enoyl)piperidin-3-yl]pyrazolo[3,4-d]pyrimidin-3-yl}-N-(4-isopropyl-3-methylphenyl)benzamide
Molecular Weight (g/mol): 523.64
Molecular Formula: C30H33N7O2
InChi Key: CGJVMKJGKFEHTL-UHFFFAOYNA-N
SMILES: CC(C)C1=C(C)C=C(NC(=O)C2=CC=CC(=C2)C2=NN(C3CCCN(C3)C(=O)C=C)C3=NC=NC(N)=C23)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results