accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14099-05-9

IUPAC Name: λ²-iron(2+) trisodium pentakis(iminomethanide) amine
Synonyms: λ²-iron(2+) trisodium pentakis(cyanide) amine
Molecular Weight (g/mol): 271.94
Molecular Formula: C5H3FeN6Na3
InChi Key: OGMPDCMTFVOHSM-UHFFFAOYSA-N
SMILES: N.[Na+].[Na+].[Na+].[Fe++].[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results