accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 141-36-6

IUPAC Name: oxirane-2,3-dicarboxylic acid
Synonyms: trans-2,3-epoxysuccinic acid
Molecular Weight (g/mol): 132.07
Molecular Formula: C4H4O5
InChi Key: DCEMCPAKSGRHCN-UHFFFAOYNA-N
SMILES: OC(=O)C1OC1C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results