Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+/-)-trans-Epoxysuccinic Acid 97.0+%, TCI America™
SDP

Catalog No. E035025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E035025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E035025G Supplier TCI America Supplier No. E035025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 141-36-6
Molecular Formula C4H4O5
Molecular Weight (g/mol) 132.071
MDL Number MFCD00070513
InChI Key DCEMCPAKSGRHCN-JCYAYHJZSA-N
Synonym (+/-)-trans-2,3-Oxiranedicarboxylic Acid
PubChem CID 440044
ChEBI CHEBI:15900
IUPAC Name (2R,3R)-oxirane-2,3-dicarboxylic acid
SMILES C1(C(O1)C(=O)O)C(=O)O

Specifications

Color White
Quantity 25 g
Formula Weight 132.07
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (+/-)-trans-Epoxysuccinic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.