accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OH3CCH3CH3HO

CAS RN 141-73-1

IUPAC Name: 4-methoxy-4-methylpentan-2-ol
Synonyms: 4-methoxy-4-methylpentan-2-ol
Molecular Weight (g/mol): 132.2
Molecular Formula: C7H16O2
InChi Key: OMSLSFZIEVFEIH-UHFFFAOYNA-N
SMILES: COC(C)(C)CC(C)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results