accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14110-64-6

IUPAC Name: (9R,12aS,13S,15S,15aS,16S,18aS)-16-benzyl-13-hydroxy-9,15-dimethyl-14-methylidene-2H,5H,6H,7H,8H,9H,10H,12aH,13H,14H,15H,15aH,16H,17H,18H-oxacyclotetradeca[3,2-e]isoindole-2,5,18-trione
Synonyms: cytochalasin A
Molecular Weight (g/mol): 477.6
Molecular Formula: C29H35NO5
InChi Key: ZMAODHOXRBLOQO-TZVKRXPSSA-N
SMILES: C[C@H]1[C@H]2[C@H](CC3=CC=CC=C3)NC(=O)[C@]22OC(=O)\C=C\C(=O)CCC[C@@H](C)C\C=C\[C@H]2[C@H](O)C1=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results