accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3SSNHN

CAS RN 141400-58-0

IUPAC Name: 2-(butan-2-yldisulfanyl)-1H-imidazole
Synonyms: 2-(sec-butyldisulfanyl)-1H-imidazole
Molecular Weight (g/mol): 188.31
Molecular Formula: C7H12N2S2
InChi Key: BPBPYQWMFCTCNG-UHFFFAOYNA-N
SMILES: CCC(C)SSC1=NC=CN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)