accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14143-32-9

IUPAC Name: 4-chloro-3-ethylphenol
Synonyms: 4-chloro-3-ethylphenol
Molecular Weight (g/mol): 156.61
Molecular Formula: C8H9ClO
InChi Key: DVKVZPIRWWREJC-UHFFFAOYSA-N
SMILES: CCC1=CC(O)=CC=C1Cl
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)
Quantity 
  • (2)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)