Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-3-ethylphenol 95.0+%, TCI America™
SDP

Catalog No. C136425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C136425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C136425G Supplier TCI America Supplier No. C136425G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 14143-32-9
Molecular Formula C8H9ClO
Molecular Weight (g/mol) 156.609
MDL Number MFCD00010822
InChI Key DVKVZPIRWWREJC-UHFFFAOYSA-N
Synonym 4-chloro-3-ethyl-phenol, phenol, 4-chloro-3-ethyl, clmep, pubchem2213, 4chloro-3-ethylphenol, 3-ethyl-4-chlorophenol, acmc-1c2gm, diisopropylidenemannofuranose, ksc183m9d, 4-chloro-3-ethylphenol
PubChem CID 84223
IUPAC Name 4-chloro-3-ethylphenol
SMILES CCC1=C(C=CC(=C1)O)Cl

Specifications

Melting Point 49°C
Color White-Yellow
Quantity 25 g
Formula Weight 156.61
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-3-ethylphenol
EINECSNumber (3)-3852
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.