Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-chloro-3-ethylphenol, 97%, Thermo Scientific™

Catalog No. AC440720250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC440720250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC440720250 Supplier Thermo Scientific Chemicals Supplier No. 440720250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

Chemical Identifiers

CAS 14143-32-9
Molecular Formula C8H9ClO
Molecular Weight (g/mol) 156.61
InChI Key DVKVZPIRWWREJC-UHFFFAOYSA-N
Synonym 4-chloro-3-ethyl-phenol, phenol, 4-chloro-3-ethyl, clmep, pubchem2213, 4chloro-3-ethylphenol, 3-ethyl-4-chlorophenol, acmc-1c2gm, diisopropylidenemannofuranose, ksc183m9d, 4-chloro-3-ethylphenol
PubChem CID 84223
IUPAC Name 4-chloro-3-ethylphenol
SMILES CCC1=C(C=CC(=C1)O)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 49.4°C to 54.3°C
Color Colorless or White to Yellow
Boiling Point 70°C to 80°C (0.2 Torr)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 25 g
Formula Weight 156.61
Percent Purity 97%
Physical Form Powder or Low Melting Solid
Chemical Name or Material 4-chloro-3-ethylphenol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 237-991-6

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.