accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OONClClH2NCl

CAS RN 14143-55-6

IUPAC Name: methyl 4-amino-3,5,6-trichloropyridine-2-carboxylate
Synonyms: methyl 4-amino-3,5,6-trichloropyridine-2-carboxylate
Molecular Weight (g/mol): 255.48
Molecular Formula: C7H5Cl3N2O2
InChi Key: RJQUHEYNLDNJLN-UHFFFAOYSA-N
SMILES: COC(=O)C1=NC(Cl)=C(Cl)C(N)=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results