Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Picloram methyl ester, Restek
SDP

Catalog No. 06719886 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-719-886 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-719-886 Supplier Restek Corporation Supplier No. 32266

Chemical Identifiers

CAS 14143-55-6
Molecular Formula C7H5Cl3N2O2
Molecular Weight (g/mol) 255.479
InChI Key RJQUHEYNLDNJLN-UHFFFAOYSA-N
PubChem CID 84224
IUPAC Name methyl 4-amino-3,5,6-trichloropyridine-2-carboxylate
SMILES COC(=O)C1=NC(=C(C(=C1Cl)N)Cl)Cl

Specifications

Quantity 1 mL
Chemical Name or Material Picloram methyl ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.