accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14150-94-8

IUPAC Name: 1-methyl-3,5-dinitro-1,2-dihydropyridin-2-one
Synonyms: 1-methyl-3,5-dinitropyridin-2-one
Molecular Weight (g/mol): 199.12
Molecular Formula: C6H5N3O5
InChi Key: QARVELJVEBLWGK-UHFFFAOYSA-N
SMILES: CN1C=C(C=C(C1=O)[N+]([O-])=O)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results