accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1416324-85-0

IUPAC Name: hexyl 3,4,5-trihydroxy-2-methoxy-6-oxo-6H-benzo[7]annulene-8-carboxylate
Synonyms: hexyl 1,2,9-trihydroxy-3-methoxy-8-oxobenzo[7]annulene-6-carboxylate
Molecular Weight (g/mol): 362.38
Molecular Formula: C19H22O7
InChi Key: UBQFTTUFRSSOHE-UHFFFAOYSA-N
SMILES: CCCCCCOC(=O)C1=CC(=O)C(O)=C2C(O)=C(O)C(OC)=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)