accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1417985-27-3

CH2FFMgBr

CAS RN 1417985-27-3

IUPAC Name: magnesium(2+) (3,5-difluorophenyl)methanide bromide
Synonyms: magnesium(2+) (3,5-difluorophenyl)methanide bromide
Molecular Weight (g/mol): 231.32
Molecular Formula: C7H5BrF2Mg
InChi Key: QAXANYLHEOPEGM-UHFFFAOYSA-M
SMILES: [Mg++].[Br-].[CH2-]C1=CC(F)=CC(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results