accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1418033-25-6

IUPAC Name: 6-{[(3,5-dimethylphenyl)formamido]oxy}-N-hydroxyhexanamide
Synonyms: 6-{[(3,5-dimethylphenyl)formamido]oxy}-N-hydroxyhexanamide
Molecular Weight (g/mol): 294.35
Molecular Formula: C15H22N2O4
InChi Key: VRYZCEONIWEUAV-UHFFFAOYSA-N
SMILES: CC1=CC(=CC(C)=C1)C(=O)NOCCCCCC(=O)NO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results