accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClO(R)OO

CAS RN 141942-53-2

IUPAC Name: (2R)-3,3-dichlorooxan-2-yl formate
Synonyms: (2R)-3,3-dichlorooxan-2-yl formate
Molecular Weight (g/mol): 199.03
Molecular Formula: C6H8Cl2O3
InChi Key: RJZSBBQFYWZFDG-RXMQYKEDSA-N
SMILES: ClC1(Cl)CCCO[C@@H]1OC=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results