CAS RN 1419949-20-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ NVP-TNKS656
Tankyrase (AKA PARP5) inhibitor; used to generate neurons from epiblast stem cells