accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CH3CCH3CH3SCH3FPFFClRu(?)(?)

CAS RN 1420299-84-8

IUPAC Name: [2',4',6'-trimethyl-3-(2,4,6-trimethylphenyl)-[1,1'-biphenyl]-2-yl]sulfanide; chlororutheniumylium; tris(4-fluorophenyl)phosphane
Synonyms: [2',4',6'-trimethyl-3-(2,4,6-trimethylphenyl)-[1,1'-biphenyl]-2-yl]sulfanide; chlororutheniumylium; tris(4-fluorophenyl)phosphane
Molecular Weight (g/mol): 798.31
Molecular Formula: C42H37ClF3PRuS
InChi Key: PCBRQBCMBTZBQC-UHFFFAOYSA-L
SMILES: Cl[Ru+].FC1=CC=C(C=C1)P(C1=CC=C(F)C=C1)C1=CC=C(F)C=C1.CC1=CC(C)=C(C(C)=C1)C1=CC=CC(=C1[S-])C1=C(C)C=C(C)C=C1C
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results