accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COSOOBr

CAS RN 1421261-59-7

IUPAC Name: methyl 3-bromobenzene-1-sulfonate
Synonyms: methyl 3-bromobenzenesulfonate
Molecular Weight (g/mol): 251.09
Molecular Formula: C7H7BrO3S
InChi Key: RBLPSOBJNBVDMD-UHFFFAOYSA-N
SMILES: COS(=O)(=O)C1=CC(Br)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results